missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2':6',2″-Terpyridine, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 89.0°C to 91.0°C |
| Boiling Point | 370.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Quantity | 250 mg |
| Formula Weight | 233.28 |
| Percent Purity | 96% |
| Chemical Name or Material | 2,2':6',2''-Terpyridine |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF exposed or concerned: |
| EINECSNumber | 214-559-5 |
RUO – Research Use Only