missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%

Catalog No. p-7052608
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH37760MD 250 mg
AAH3776003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH37760MD Supplier Thermo Scientific Chemicals Supplier No. H37760MD

CAS: 148332-36-9 | C16H11N3O2 | 277.28 g/mol

2,2':6',2″-Terpyridine-4'-carboxylic acid is used as dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:6′,2′′-Terpyridine-4′-carboxylic acid is used as dyestuff intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 148332-36-9
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.28
MDL Number MFCD04114290
InChI Key ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
PubChem CID 2762749
IUPAC Name 2,6-dipyridin-2-ylpyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 280°C to 282°C
Color White to Cream
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 277.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,2':6',2''-Terpyridine-4'-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only