missing translation for 'onlineSavingsMsg'
Learn More

2,2,5-Trimethylhexane 99.0+%, TCI America™

Catalog No. p-7136303
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04755ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T04755ML Supplier TCI America Supplier No. T04755ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3522-94-9
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00026525
InChI Key HHOSMYBYIHNXNO-UHFFFAOYSA-N
PubChem CID 19041
IUPAC Name 2,2,5-trimethylhexane
SMILES CC(C)CCC(C)(C)C

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,5-Trimethylhexane
EINECSNumber (2)-0009
RTECSNumber MO4630000
TSCA No