accessibility menu, dialog, popup

UserName

CAS RN 3522-94-9

H3CCH3CH3CH3H3C

CAS RN 3522-94-9

IUPAC Name: 2,2,5-trimethylhexane
Synonyms: 2,2,5-trimethylhexane
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: HHOSMYBYIHNXNO-UHFFFAOYSA-N
SMILES: CC(C)CCC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results