Learn More
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%
CAS: 19805-75-5 | C9H16N2O | 168.24 g/mol
$94.81 - $287.74
Chemical Identifiers
| CAS | 19805-75-5 |
|---|---|
| Molecular Formula | C9H16N2O |
| Molecular Weight (g/mol) | 168.24 |
| MDL Number | MFCD00041846 |
| InChI Key | ACFYUJLIWIDSFM-UHFFFAOYSA-N |
| Synonym | 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide, 2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide, 1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl, 2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide, 3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline, maybridge3_000440, acmc-1c3ma, 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline, 3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline |
| PubChem CID | 88255 |
| IUPAC Name | 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide |
| SMILES | CC1(C=C(C(N1)(C)C)C(=O)N)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1829403
|
Thermo Scientific Chemicals
A1829403 |
1 g |
Each for $94.81
|
|
|||||
|
AAA1829406
|
Thermo Scientific Chemicals
A1829406 |
5 g |
Each for $287.74
|
|
|||||
Description
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 19805-75-5 | |
| 168.24 | |
| ACFYUJLIWIDSFM-UHFFFAOYSA-N | |
| 88255 | |
| CC1(C=C(C(N1)(C)C)C(=O)N)C |
| C9H16N2O | |
| MFCD00041846 | |
| 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide, 2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide, 1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl, 2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide, 3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline, maybridge3_000440, acmc-1c3ma, 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline, 3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline | |
| 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide |
Specifications
| 19805-75-5 | |
| C9H16N2O | |
| 1 g | |
| Slightly soluble in water. | |
| CC1(C=C(C(N1)(C)C)C(=O)N)C | |
| 168.24 | |
| 168.24 | |
| 2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide |
| 179°C to 183°C | |
| MFCD00041846 | |
| 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide, 2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide, 1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl, 2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide, 3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline, maybridge3_000440, acmc-1c3ma, 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline, 3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline | |
| ACFYUJLIWIDSFM-UHFFFAOYSA-N | |
| 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide | |
| 88255 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 243-331-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only