CAS RN 19805-75-5
IUPAC Name:
3-carbamoyl-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrol-1-ium
Synonyms:
3-carbamoyl-2,2,5,5-tetramethyl-1H-pyrrol-1-ium
Molecular Weight (g/mol):
169.25
Molecular Formula:
C9H17N2O
InChi Key:
ACFYUJLIWIDSFM-UHFFFAOYSA-O
SMILES:
CC1(C)[NH2+]C(C)(C)C(=C1)C(N)=O
1
–
1
of
1
results
1
Promotions Available
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%
CAS: 19805-75-5 Molecular Formula: C9H16N2O Molecular Weight (g/mol): 168.24 MDL Number: MFCD00041846 InChI Key: ACFYUJLIWIDSFM-UHFFFAOYSA-N Synonym: 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide,2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide,1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl,2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide,3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline,maybridge3_000440,acmc-1c3ma,3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline,3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline PubChem CID: 88255 IUPAC Name: 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide SMILES: CC1(C=C(C(N1)(C)C)C(=O)N)C