missing translation for 'onlineSavingsMsg'
Learn More

2,2':5',2″-Terthiophene (purified by sublimation) 98.0+%, TCI America™

Catalog No. T11961G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11961G 1 g
T11965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T11961G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T11961G

Chemical Identifiers

CAS 1081-34-1
Molecular Formula C12H8S3
Molecular Weight (g/mol) 248.376
MDL Number MFCD00012167
InChI Key KXSFECAJUBPPFE-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienyl
PubChem CID 65067
ChEBI CHEBI:10335
IUPAC Name 2,5-dithiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3

Specifications

Melting Point 95°C
Quantity 1 g
Formula Weight 248.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′:5′,2′′-Terthiophene (purified by sublimation)
RTECSNumber WZ9717750
TSCA No