accessibility menu, dialog, popup

UserName

CAS RN 1081-34-1

IUPAC Name: 5-(thiophen-2-yl)-2,2'-bithiophene
Synonyms: terthiophene α-terthienyl 2,5-thiophenyl-thiophene α-terthiophene
Molecular Weight (g/mol): 248.38
Molecular Formula: C12H8S3
InChi Key: KXSFECAJUBPPFE-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(S1)C1=CC=CS1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)