missing translation for 'onlineSavingsMsg'
Learn More

2,2':5',2″-Terthiophene-5,5″-dicarboxaldehyde 98.0+%, TCI America™

Catalog No. T26391G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26391G Supplier TCI America Supplier No. T26391G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13130-50-2
Molecular Formula C14H8O2S3
Molecular Weight (g/mol) 304.396
InChI Key YAEGPDBHSBKYRW-UHFFFAOYSA-N
Synonym 5,5′C′C-Diformyl-2,2′C:5′C,2′C′C-terthiophene
PubChem CID 4389805
IUPAC Name 5-[5-(5-formylthiophen-2-yl)thiophen-2-yl]thiophene-2-carbaldehyde
SMILES C1=C(SC(=C1)C2=CC=C(S2)C3=CC=C(S3)C=O)C=O

Specifications

Melting Point 222°C
Quantity 1 g
Formula Weight 304.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′:5′,2′′-Terthiophene-5,5′′-dicarboxaldehyde