accessibility menu, dialog, popup

UserName

SSSOO

CAS RN 13130-50-2

IUPAC Name: 5'-(5-formylthiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Synonyms: 5'-(5-formylthiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Molecular Weight (g/mol): 304.4
Molecular Formula: C14H8O2S3
InChi Key: YAEGPDBHSBKYRW-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results