missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99+%, for spectroscopy ACS

CAS: 540-84-1 | C8H18 | 114.232 g/mol

Supplier:  Thermo Scientific Chemicals 421980025

 View more versions of this product

Catalog No. AC421980025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

2, 2, 4-Trimethylpentane
540-84-1
99.0
0.6900g/mL
2.5 L
Glass Bottle
1.3915
99+%
0.0003 meq/g max.
(CH3)2CHCH2C(CH3)3
0.51 mPa.s (22°C)
Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil
CC(C)CC(C)(C)C
114.232
CHEBI:62805
≥99%
-107.0°C
0.01 max. at 250nm, 0.01 max. at 400nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 1 max. at 210nm
100.0
98.0°C to 99.0°C
−12°C
Liquid
0.001% max.
0.69
C8H18
MFCD00008943
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23
ACS

Chemical Identifiers

540-84-1
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane
C8H18
MFCD00008943
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-759-1

RTECSNumber : SA3320000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only