missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, For pesticide residue analysis, Solstice

Catalog No. 6000109
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000109 4 x 4 L
6000865 1 L
6000866 2.5 L
6000867 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000109 Supplier Solstice Supplier No. 344994X4L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C (literature)
CAS Max % 100.0000%
Density 0.692 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point −12°C
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2C(CH3)3
UN Number UN1262
Boiling Point 99°C
Formula Weight 114.23
Percent Purity >99%
Grade Pesticide
Chemical Name or Material 2,2,4-Trimethylpentane, For pesticide residue analysis
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes
EINECSNumber 208-759-1
Recommended Storage Room Temp