missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, CHROMASOLV™ Plus, for HPLC,≥99.5%, Solstice

Catalog No. 60048245
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048245 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048245 Supplier Solstice Supplier No. 6504394X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym Isooctane
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

CAS Max % 100.00
Density 0.69 g/cm3
Quantity 4 x 4 L
Flash Point -8°C
Packaging Glass Bottle
Physical Form Liquid
Beilstein 1696876
Boiling Point 99°C
Grade HPLC
Hazard Category Danger
Hazard Statement H225+H304+H315+H336
Precautionary Statement P280+P271+P264+P261+P243+P242+P241+P240+P233+P210
RTECSNumber 208-759-1
Recommended Storage Room Temperature