missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019809
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019809 5 mL
111019005 10 mL
111018171 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111019809 Supplier Merck Emd Millipore Supplier No. 590305ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,2,4-Trimethylpentane, a branched alkane, having a low tendency to auto-ignite, is used as a reference in determining the octane number of gasoline.

Specifications

CAS 540-84-1
Vapor Pressure 41 mmHg (21°C)
Vapor Density 3.9 (vs. air)
Density 0.692 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2C(CH3)3
MDL Number MFCD00008943
Melting Point -107°C
Boiling Point 98°C to 99°C
Synonym Isooctane
Quantity 5 mL
Formula Weight 114.23
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less