missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99%, Spectrum™ Chemical

Catalog No. 18608159
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608159 500 mL Amber Glass Bottle
18608164 1 L Amber Glass Bottle
18608165 4 L Amber Glass Bottle
18608157 20 L Steel Can
18608158 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608159 Supplier Spectrum Chemical Mfg Cor Supplier No. T1125500MLGL

Chemical Identifiers

CAS 540-84-1
Molecular Weight (g/mol) 114.23

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Boiling Point 99.238°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material 2,2,4-Trimethylpentane

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.