missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ≥99%, Reagent Grade, Honeywell™
Reagent Grade,≥99%
$286.98 - $286.98
Chemical Identifiers
| CAS | 540-84-1 |
|---|---|
| Molecular Formula | C8H18 |
| Molecular Weight (g/mol) | 114.232 |
| MDL Number | MFCD00008943 |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
| PubChem CID | 10907 |
| ChEBI | CHEBI:62805 |
| IUPAC Name | 2,2,4-trimethylpentane |
| SMILES | CC(C)CC(C)(C)C |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 540-84-1 | |
| 114.232 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 10907 | |
| 2,2,4-trimethylpentane |
| C8H18 | |
| MFCD00008943 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CHEBI:62805 | |
| CC(C)CC(C)(C)C |
Specifications
| -107°C | |
| Colorless | |
| 98°C to 99°C | |
| 0.06 | |
| 745°F | |
| Liquid | |
| (CH3)2CHCH2C(CH3)3 | |
| 41mmHg (21°C) | |
| 1696876 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 2,2,4-trimethylpentane | |
| 10907 | |
| 114.23g/mol | |
| ReagentPlus |
| 540-84-1 | |
| 0.692g/mL at 25°C | |
| 2.5 L | |
| −12°C (10.4°F) | |
| Glass Bottle | |
| C8H18 | |
| MFCD00008943 | |
| UN1262 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CC(C)CC(C)(C)C | |
| 114.232 | |
| CHEBI:62805 | |
| ≥99% | |
| 2,2,4-Trimethylpentane |
Safety and Handling
P210-P273-P301 + P310-P331-P370 + P378-P501
H225-H304-H315-H336-H410
DOTInformation : Class 3 - PG 2 - Octanes
EINECSNumber : 208-759-1
RTECSNumber : SA3320000