Learn More
2,2,4-Trimethylpentane, 99.5%, for HPLC
For HPLC | CAS: 540-84-1 | C8H18 | 114.232 g/mol
Supplier: Thermo Scientific Chemicals 268810025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
| 2, 2, 4-Trimethylpentane | |
| 540-84-1 | |
| Colorless to Yellow | |
| 100.0 | |
| 0.6900g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.3915 | |
| 99.5% | |
| C8H18 | |
| MFCD00008943 | |
| 0.01% Max. | |
| 15, 5239 | |
| Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil | |
| CC(C)CC(C)(C)C | |
| 114.232 | |
| CHEBI:62805 | |
| 99.5% |
| -107.0°C | |
| (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 240nm, 0.005 max. at 250nm, 0.02 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.4 max. at 200nm | |
| 99.4 | |
| 0.0002 meq/g max. | |
| 98.0°C to 99.0°C | |
| −12°C | |
| Liquid | |
| 0.0002 % max. | |
| 0.69 | |
| (CH3)2CHCH2C(CH3)3 | |
| 0.51 mPa.s (22°C) | |
| 01, IV, 439 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 2,2,4-trimethylpentane | |
| 10907 | |
| 114.23 | |
| HPLC |
Chemical Identifiers
| 540-84-1 | |
| 114.232 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 10907 | |
| 2,2,4-trimethylpentane |
| C8H18 | |
| MFCD00008943 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CHEBI:62805 | |
| CC(C)CC(C)(C)C |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-759-1
RTECSNumber : SA3320000
TSCA : TSCA
RUO – Research Use Only