missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99.5%, for HPLC
GREENER_CHOICE

Catalog No. AC268810010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268810010 1 L Glass Bottle
AC268810025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268810010 Supplier Thermo Scientific Chemicals Supplier No. 268810010
Get 1 Extra Chemical When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For HPLC, CAS: 540-84-1, C8H18, 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107.0°C
Absorbance 1 cm cell vs HPLC-grade water, 1 cm cell vs HPLC-grade water at 0 nm, 0.005 max. at 240nm, 0.005 max. at 250nm, 0.02 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.4 max. at 200nm
Color Colorless to Yellow
CAS Min % 99.4
CAS Max % 100.0
Acidity 0.0002 meq/g max.
Density 0.6900g/mL
Quantity 1 L
Flash Point -12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3915
Residue after Evaporation 0.0002 % max.
Specific Gravity 0.69
Assay Percent Range 99.5%
Linear Formula (CH3)2CHCH2C(CH3)3
Viscosity 0.51 mPa.s (22°C)
Water 0.01% Max.
Beilstein 01, IV, 439
Merck Index 15, 5239
Green Features ACT Ecolabel (Gold)
Boiling Point 98.0°C to 99.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil
Formula Weight 114.23
Percent Purity 99.5%
Grade HPLC
Chemical Name or Material 2, 2, 4-Trimethylpentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-759-1
RTECSNumber SA3320000
TSCA TSCA

RUO – Research Use Only