missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Dinitrobenzyl)pyridine 98.0+%, TCI America™

Catalog No. D10951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10951G 1 g
D10955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10951G Supplier TCI America Supplier No. D10951G

Chemical Identifiers

CAS 1151-97-9
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.221
MDL Number MFCD00014632
InChI Key KKFNJVINGIUTIH-UHFFFAOYSA-N
PubChem CID 70854
IUPAC Name 2-[(2,4-dinitrophenyl)methyl]pyridine
SMILES C1=CC=NC(=C1)CC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 90°C
Color Yellow
Quantity 1 g
Formula Weight 259.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,4-Dinitrobenzyl)pyridine
EINECSNumber (5)-3721
TSCA Yes