missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,6,6-Pentamethylheptane

Catalog No. p-7214403
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462050050 5 mL
AC462050250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462050050 Supplier Thermo Scientific Chemicals Supplier No. 462050050
Only null left

CAS: 13475-82-6 | C12H26 | 170.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
IUPAC Name 2,2,4,6,6-pentamethylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C

Specifications

Density 0.7463 g/mL
Boiling Point 170.0°C
Flash Point 37°C
Linear Formula [CH3C(CH3)2CH2]2CHCH3
Quantity 5 mL
Specific Gravity 0.7463
Formula Weight 170.33
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable liquid and vapor.

May be fatal if swallowed and enters airways.

May cause long lasting harmful effects to aquatic life.

Repeated exposure may cause skin dryness or cracking.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Do NOT induce vomiting.

Store in a well-ventilated place.

Keep container tightly closed.

EINECSNumber 236-757-0

RUO – Research Use Only