accessibility menu, dialog, popup

UserName

H3CH3CCH3H3CCH3CH3H3C

CAS RN 13475-82-6

IUPAC Name: 2,2,4,6,6-pentamethylheptane
Synonyms: 2,2,4,6,6-pentamethylheptane
Molecular Weight (g/mol): 170.34
Molecular Formula: C12H26
InChi Key: VKPSKYDESGTTFR-UHFFFAOYSA-N
SMILES: CC(CC(C)(C)C)CC(C)(C)C
Boiling Point 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)