missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,6,6-Pentamethylheptane 98.0+%, TCI America™

Catalog No. p-7134391
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00475ML 5 mL
P004725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00475ML Supplier TCI America Supplier No. P00475ML
Only null left

Chemical Identifiers

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00026334
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
PubChem CID 26058
ChEBI CHEBI:77509
IUPAC Name 2,2,4,6,6-pentamethylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 5 mL
UN Number 2286
Synonym heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane
Formula Weight 170.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane
EINECSNumber (2)-0010
TSCA Yes