missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethyl-3-pentanone Imine 98.0+%, TCI America™

Catalog No. T29891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29891G 1 g
T29895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29891G Supplier TCI America Supplier No. T29891G

Chemical Identifiers

CAS 29097-52-7
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.258
MDL Number MFCD00216566
InChI Key VCMKYJMEMATQPE-UHFFFAOYSA-N
Synonym 2,2,4,4-Tetramethyl-3-pentanimine, Di-tert-butylmethanimine
PubChem CID 2755632
IUPAC Name 2,2,4,4-tetramethylpentan-3-imine
SMILES CC(C)(C)C(=N)C(C)(C)C

Specifications

Color Colorless
Boiling Point 164°C
Quantity 1 g
UN Number 1993
Formula Weight 141.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,4,4-Tetramethyl-3-pentanone Imine
TSCA No
Recommended Storage Refrigerator