accessibility menu, dialog, popup

UserName

CAS RN 29097-52-7

IUPAC Name: 2,2,4,4-tetramethylpentan-3-imine
Synonyms: 2,2,4,4-tetramethylpentan-3-imine
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: VCMKYJMEMATQPE-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=N)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results