missing translation for 'onlineSavingsMsg'
Learn More

2,2,3-Trimethylpentane 99.0+%, TCI America™

Catalog No. T06891ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06891ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06891ML Supplier TCI America Supplier No. T06891ML
Only null left

Chemical Identifiers

CAS 564-02-3
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00059241
InChI Key XTDQDBVBDLYELW-UHFFFAOYSA-N
PubChem CID 11255
IUPAC Name 2,2,3-trimethylpentane
SMILES CCC(C)C(C)(C)C

Specifications

Color Colorless
Boiling Point 110°C
Quantity 1 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3-Trimethylpentane
EINECSNumber (2)-0008
TSCA No