accessibility menu, dialog, popup

UserName

CAS RN 564-02-3

CH3CH3CH3CH3H3C

CAS RN 564-02-3

IUPAC Name: 2,2,3-trimethylpentane
Synonyms: 2,2,3-trimethylpentane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: XTDQDBVBDLYELW-UHFFFAOYNA-N
SMILES: CCC(C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results