missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluoro-1-butanol, 95%

Catalog No. p-7025212
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2058706 5 g
AAB2058714 25 g
AAB2058722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2058706 Supplier Thermo Scientific Chemicals Supplier No. B2058706
Only null left

CAS: 382-31-0 | C4H4F6O | 182.065 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 382-31-0
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00042309
InChI Key LVFXLZRISXUAIL-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluoro-1-butanol, 1h,1h,3h-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol-1, 2,2,3,4,4,4-hexafluoro-butan-1-ol, 1-butanol, 2,2,3,4,4,4-hexafluoro, 1h,1h,3h-perfluorobutan-1-ol, acmc-209iys, cf3cfhcf2ch2oh, 2,2,3,4,4,4-hexafluorobutyl alcohol
PubChem CID 533990
IUPAC Name 2,2,3,4,4,4-hexafluorobutan-1-ol
SMILES C(C(C(C(F)(F)F)F)(F)F)O

Specifications

Density 1.64
Boiling Point 114°C to 116°C
Flash Point 51°C (123°F)
Refractive Index 1.312
Quantity 5 g
UN Number UN1993
Beilstein 1753872
Formula Weight 182.07
Percent Purity 95%
Chemical Name or Material 2,2,3,4,4,4-Hexafluoro-1-butanol
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-842-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only