accessibility menu, dialog, popup

UserName

CAS RN 382-31-0

IUPAC Name: 2,2,3,4,4,4-hexafluorobutan-1-ol
Synonyms: 2,2,3,4,4,4-hexafluorobutan-1-ol
Molecular Weight (g/mol): 182.07
Molecular Formula: C4H4F6O
InChi Key: LVFXLZRISXUAIL-UHFFFAOYNA-N
SMILES: OCC(F)(F)C(F)C(F)(F)F
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)