missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3-Tetramethylcyclopropanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America T235025G
Specifications
| 2,2,3,3-Tetramethylcyclopropanecarboxylic Acid | |
| 120°C | |
| C8H14O2 | |
| 25 g | |
| SFHVXKNMCGSLAR-UHFFFAOYSA-N | |
| 2,2,3,3-tetramethylcyclopropane-1-carboxylic acid | |
| 152115 | |
| 142.20 | |
| Crystalline Powder |
| 15641-58-4 | |
| White | |
| MFCD00011538 | |
| 2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid | |
| CC1(C(C1(C)C)C(=O)O)C | |
| 142.198 | |
| CHEBI:39355 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 15641-58-4 | |
| 142.198 | |
| SFHVXKNMCGSLAR-UHFFFAOYSA-N | |
| 152115 | |
| 2,2,3,3-tetramethylcyclopropane-1-carboxylic acid |
| C8H14O2 | |
| MFCD00011538 | |
| 2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid | |
| CHEBI:39355 | |
| CC1(C(C1(C)C)C(=O)O)C |
Safety and Handling
TSCA : No