accessibility menu, dialog, popup

UserName

CAS RN 15641-58-4

CH3H3COHOH3CH3C

CAS RN 15641-58-4

IUPAC Name: tetramethylcyclopropane-1-carboxylic acid
Synonyms: tetramethylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: SFHVXKNMCGSLAR-UHFFFAOYSA-N
SMILES: CC1(C)C(C(O)=O)C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results