missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoropropyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. T22405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22405G 5 g
T224025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22405G Supplier TCI America Supplier No. T22405G

Chemical Identifiers

CAS 7383-71-3
Molecular Formula C6H6F4O2
Molecular Weight (g/mol) 186.11
MDL Number MFCD00042380
InChI Key VHJHZYSXJKREEE-UHFFFAOYSA-N
Synonym Acrylic Acid 2,2,3,3-Tetrafluoropropyl Ester
PubChem CID 81865
IUPAC Name 2,2,3,3-tetrafluoropropyl prop-2-enoate
SMILES FC(F)C(F)(F)COC(=O)C=C

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 g
UN Number 3272
Formula Weight 186.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl Acrylate (stabilized with MEHQ)
TSCA No
Recommended Storage Refrigerator