accessibility menu, dialog, popup

UserName

FFFFOOH2C

CAS RN 7383-71-3

IUPAC Name: 2,2,3,3-tetrafluoropropyl prop-2-enoate
Synonyms: 2,2,3,3-tetrafluoropropyl prop-2-enoate
Molecular Weight (g/mol): 186.11
Molecular Formula: C6H6F4O2
InChi Key: VHJHZYSXJKREEE-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)COC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results