missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',3,3',5,5',6,6'-Octafluorobiphenyl 97.0+%, TCI America™
$55.92 - $165.90
Chemical Identifiers
| CAS | 3883-86-1 |
|---|---|
| Molecular Formula | C12H2F8 |
| Molecular Weight (g/mol) | 298.14 |
| MDL Number | MFCD00000312 |
| InChI Key | QWCHHUZAAGRHDB-UHFFFAOYSA-N |
| Synonym | 4H,4′CH-Octafluorobiphenyl |
| PubChem CID | 77492 |
| IUPAC Name | 2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl |
| SMILES | FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F |
Chemical Identifiers
| 3883-86-1 | |
| 298.14 | |
| QWCHHUZAAGRHDB-UHFFFAOYSA-N | |
| 77492 | |
| FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F |
| C12H2F8 | |
| MFCD00000312 | |
| 4H,4′CH-Octafluorobiphenyl | |
| 2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl |
Specifications
| 3883-86-1 | |
| C12H2F8 | |
| White | |
| 4H,4′CH-Octafluorobiphenyl | |
| FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F | |
| 298.14 | |
| 77492 | |
| ≥97.0% (GC) |
| Powder/Crystals | |
| MFCD00000312 | |
| 85°C | |
| QWCHHUZAAGRHDB-UHFFFAOYSA-N | |
| 2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl | |
| 1 g | |
| 298.14 | |
| 2,2′,3,3′,5,5′,6,6′-Octafluorobiphenyl |
Safety and Handling
TSCA : Yes