missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Staphylococcus aureus BioParticles™ Opsonizing Reagent

Catalog No. 502723477
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502723477 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502723477 Supplier Invitrogen™ Supplier No. LSS2860

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

The Staphylococcus aureus (Wood strain without protein A) BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

The Staphylococcus aureus (Wood strain without protein A) BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies and specifically designed to enhance the uptake of the S. aureus BioParticles™ conjugates.

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 13485

Compliance

GHS/OSHA Complaint

Specifications

Product Line BioParticles
Shipping Condition Room Temperature
Molecular Formula C12H2F8
MDL Number MFCD00000312
For Use With (Equipment) Fluorescence Microscope
InChI Key QWCHHUZAAGRHDB-UHFFFAOYSA-N
SMILES FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F
IUPAC Name 2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
Molecular Weight (g/mol) 298.14
Quantity 1 unit
Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

For Research Use Only. Not for use in diagnostic procedures.