missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4-Hexafluoro-1,5-pentanediol 98.0+%, TCI America™

Catalog No. p-7130704
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07461G 1 g
H07465G 5 g
H074625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H07461G Supplier TCI America Supplier No. H07461G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 376-90-9
Molecular Formula C5H6F6O2
Molecular Weight (g/mol) 212.091
MDL Number MFCD00042434
InChI Key IELVMUPSWDZWSD-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4-hexafluoro-1,5-pentanediol, hexafluoroamylene glycol, 1,5-pentanediol, 2,2,3,3,4,4-hexafluoro, hexafluoro-1,5-pentanediol, 2,2,3,3,4,4-hexafluoropentanediol, 2,2,3,3,4,4-hexafluoropentan-1,5-diol, 1,5-dihydroxy-2,2,3,3,4,4-hexafluoropentane, acmc-209iuy, ielvmupswdzwsd-uhfffaoysa, 2,3,3,4,4-hexafluoropentanediol
PubChem CID 67831
IUPAC Name 2,2,3,3,4,4-hexafluoropentane-1,5-diol
SMILES C(C(C(C(CO)(F)F)(F)F)(F)F)O

Specifications

Melting Point 79°C
Color White
Boiling Point 112°C
Quantity 1 g
Formula Weight 212.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,4,4-Hexafluoro-1,5-pentanediol
RTECSNumber SA0750000
TSCA Yes