accessibility menu, dialog, popup

UserName

OHFFFFFFHO

CAS RN 376-90-9

IUPAC Name: 2,2,3,3,4,4-hexafluoropentane-1,5-diol
Synonyms: 2,2,3,3,4,4-hexafluoropentane-1,5-diol
Molecular Weight (g/mol): 212.09
Molecular Formula: C5H6F6O2
InChi Key: IELVMUPSWDZWSD-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)CO
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)