Learn More
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%
CAS: 355-80-6 | C5H4F8O | 232.07 g/mol
Supplier: Thermo Scientific Chemicals B2010822
Description
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.
Solubility
Insoluble in water.
Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
Specifications
| 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol | |
| <-50°C | |
| 141°C to 142°C | |
| C5H4F8O | |
| MFCD00039631 | |
| 1773494 | |
| 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol | |
| JUGSKHLZINSXPQ-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5-octafluoropentan-1-ol | |
| 9641 | |
| 98% |
| 355-80-6 | |
| 1.667 | |
| 75°C (167°F) | |
| 1.318 | |
| 100 g | |
| 14,6746 | |
| Insoluble in water. | |
| OCC(F)(F)C(F)(F)C(F)(F)C(F)F | |
| 232.07 | |
| 232.07 |
Chemical Identifiers
| 355-80-6 | |
| 232.07 | |
| JUGSKHLZINSXPQ-UHFFFAOYSA-N | |
| 9641 | |
| OCC(F)(F)C(F)(F)C(F)(F)C(F)F |
| C5H4F8O | |
| MFCD00039631 | |
| 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol | |
| 2,2,3,3,4,4,5,5-octafluoropentan-1-ol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 206-593-4
RTECSNumber : SA7650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only