missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%

Catalog No. p-7024866
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2010814 25 g
AAB2010822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2010814 Supplier Thermo Scientific Chemicals Supplier No. B2010814
Only null left

CAS: 355-80-6 | C5H4F8O | 232.07 g/mol

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

Solubility
Insoluble in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.

Chemical Identifiers

CAS 355-80-6
Molecular Formula C5H4F8O
Molecular Weight (g/mol) 232.07
MDL Number MFCD00039631
InChI Key JUGSKHLZINSXPQ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
PubChem CID 9641
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Melting Point <-50°C
Density 1.667
Boiling Point 141°C to 142°C
Flash Point 75°C (167°F)
Refractive Index 1.318
Quantity 25 g
Beilstein 1773494
Merck Index 14,6746
Solubility Information Insoluble in water.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-593-4
RTECSNumber SA7650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only