missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile 98.0+%, TCI America™

Supplier:  TCI America D522325G

 View more versions of this product

Catalog No. D522325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile
Yellow
C8F13NO
25 g
FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C#N
373.07
373.07
Liquid
120903-40-4
110°C
MFCD30721943
QCWQPRUOTXVNSB-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6-decafluoro-6-[(1,2,2-trifluoroethenyl)oxy]hexanenitrile
14172634
≥98.0% (GC)

Chemical Identifiers

120903-40-4
373.07
QCWQPRUOTXVNSB-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6-decafluoro-6-[(1,2,2-trifluoroethenyl)oxy]hexanenitrile
C8F13NO
MFCD30721943
14172634
FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions