accessibility menu, dialog, popup

UserName

CAS RN 120903-40-4

NOFFFFFFFFFFFFF

CAS RN 120903-40-4

IUPAC Name: 2,2,3,3,4,4,5,5,6,6-decafluoro-6-[(1,2,2-trifluoroethenyl)oxy]hexanenitrile
Synonyms: 2,2,3,3,4,4,5,5,6,6-decafluoro-6-[(1,2,2-trifluoroethenyl)oxy]hexanenitrile
Molecular Weight (g/mol): 373.07
Molecular Formula: C8F13NO
InChi Key: QCWQPRUOTXVNSB-UHFFFAOYSA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results