missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile 98.0+%, TCI America™

Catalog No. D52235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52235G 5 g
D522325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52235G Supplier TCI America Supplier No. D52235G
Only null left

Chemical Identifiers

CAS 120903-40-4
Molecular Formula C8F13NO
Molecular Weight (g/mol) 373.07
MDL Number MFCD30721943
InChI Key QCWQPRUOTXVNSB-UHFFFAOYSA-N
PubChem CID 14172634
IUPAC Name 2,2,3,3,4,4,5,5,6,6-decafluoro-6-[(1,2,2-trifluoroethenyl)oxy]hexanenitrile
SMILES FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C#N

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 373.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile
TSCA No
Recommended Storage Refrigerator