missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate (stabilized with TBC) 97.0+%, TCI America™

$72.70 - $221.83

Chemical Identifiers

CAS 2261-99-6
Molecular Formula C11H8F12O2
Molecular Weight (g/mol) 400.164
MDL Number MFCD00080658
InChI Key YJKHMSPWWGBKTN-UHFFFAOYSA-N
Synonym Methacrylic Acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Ester
PubChem CID 102236
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D49905G
View Documents
TCI America
D49905G
5 g N/A
Only null left
 
D499025G
View Documents
TCI America
D499025G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2261-99-6
400.164
YJKHMSPWWGBKTN-UHFFFAOYSA-N
102236
CC(=C)C(=O)OCC(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C11H8F12O2
MFCD00080658
Methacrylic Acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Ester
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate

Specifications

2261-99-6
112°C
MFCD00080658
Methacrylic Acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Ester
CC(=C)C(=O)OCC(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
400.164
400.16
Liquid
Yellow
C11H8F12O2
5 g
YJKHMSPWWGBKTN-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate
102236
≥97.0% (GC)
2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate (stabilized with TBC)
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'