missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-butanol 98.0+%, TCI America™

Catalog No. p-7130602
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05485G 5 g
H054825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05485G Supplier TCI America Supplier No. H05485G
Only null left

Chemical Identifiers

CAS 375-01-9
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.06
MDL Number MFCD00004674
InChI Key WXJFKAZDSQLPBX-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
PubChem CID 9776
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
UN Number 1987
Formula Weight 200.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-butanol