accessibility menu, dialog, popup

UserName

HOFFFFFFF

CAS RN 375-01-9

IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutan-1-ol
Synonyms: 2,2,3,3,4,4,4-heptafluorobutan-1-ol
Molecular Weight (g/mol): 200.06
Molecular Formula: C4H3F7O
InChi Key: WXJFKAZDSQLPBX-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)