missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-(2-Vinylanthracene-9,10-diylidene)dimalononitrile 98.0+%, TCI America™
$216.24 - $646.83
Chemical Identifiers
| CAS | 1612793-07-3 |
|---|---|
| Molecular Formula | C22H10N4 |
| Molecular Weight (g/mol) | 330.35 |
| InChI Key | SUTLXPZYDBOIRV-UHFFFAOYSA-N |
| Synonym | 9,10-Bis(dicyanomethylene)-2-vinylanthracene |
| PubChem CID | 91972128 |
| IUPAC Name | 2-[10-(dicyanomethylidene)-3-ethenylanthracen-9-ylidene]propanedinitrile |
| SMILES | C=CC1=CC2=C(C=C1)C(=C(C#N)C#N)C3=CC=CC=C3C2=C(C#N)C#N |
Chemical Identifiers
| 1612793-07-3 | |
| 330.35 | |
| 9,10-Bis(dicyanomethylene)-2-vinylanthracene | |
| 2-[10-(dicyanomethylidene)-3-ethenylanthracen-9-ylidene]propanedinitrile |
| C22H10N4 | |
| SUTLXPZYDBOIRV-UHFFFAOYSA-N | |
| 91972128 | |
| C=CC1=CC2=C(C=C1)C(=C(C#N)C#N)C3=CC=CC=C3C2=C(C#N)C#N |
Specifications
| 1612793-07-3 | |
| 1 g | |
| 9,10-Bis(dicyanomethylene)-2-vinylanthracene | |
| C=CC1=CC2=C(C=C1)C(=C(C#N)C#N)C3=CC=CC=C3C2=C(C#N)C#N | |
| 330.35 | |
| 330.35 | |
| Crystalline Powder |
| C22H10N4 | |
| 3439 | |
| SUTLXPZYDBOIRV-UHFFFAOYSA-N | |
| 2-[10-(dicyanomethylidene)-3-ethenylanthracen-9-ylidene]propanedinitrile | |
| 91972128 | |
| ≥98.0% (HPLC) | |
| 2,2′-(2-Vinylanthracene-9,10-diylidene)dimalononitrile |
Safety and Handling
TSCA : No