missing translation for 'onlineSavingsMsg'
Learn More

2,2'-(2-Vinylanthracene-9,10-diylidene)bis(1,3-dithiole) 98.0+%, TCI America™

Catalog No. p-7137937
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0136100MG 100 mg
V0136500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0136100MG Supplier TCI America Supplier No. V0136100MG
Only null left

Chemical Identifiers

CAS 736998-56-4
Molecular Formula C22H14S4
Molecular Weight (g/mol) 406.594
InChI Key PVHQRJPHCJRMKB-UHFFFAOYSA-N
Synonym 9,10-Bis(1,3-dithiol-2-ylidene)-2-vinylanthracene
PubChem CID 11384232
IUPAC Name 2-[10-(1,3-dithiol-2-ylidene)-2-ethenylanthracen-9-ylidene]-1,3-dithiole
SMILES C=CC1=CC2=C(C=C1)C(=C3SC=CS3)C4=CC=CC=C4C2=C5SC=CS5

Specifications

Quantity 100 mg
Formula Weight 406.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-(2-Vinylanthracene-9,10-diylidene)bis(1,3-dithiole)