missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Triphenylacetophenone 99.0+%, TCI America™

Catalog No. T12075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12075G 5 g
T120725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12075G Supplier TCI America Supplier No. T12075G
Only null left

Chemical Identifiers

CAS 466-37-5
Molecular Formula C26H20O
Molecular Weight (g/mol) 348.445
MDL Number MFCD00004762
InChI Key CFBBKHROQRFCNZ-UHFFFAOYSA-N
Synonym 2,2,2-triphenylacetophenone, tetraphenylethanone, benzopinacolone, ethanone, tetraphenyl, phenyl trityl ketone, acetophenone, 2,2,2-triphenyl, 1,2,2,2-tetraphenylethan-1-one, ss-benzpinakolin, beta-benzopinacolone, .beta.-benzopinacolone
PubChem CID 68049
IUPAC Name 1,2,2,2-tetraphenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 184°C
Color White
Quantity 5 g
Formula Weight 348.45
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Triphenylacetophenone