missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylthioacetamide, 97%

Catalog No. AAH3209203
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3209203 1 g
AAH3209209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3209203 Supplier Thermo Scientific Chemicals Supplier No. H3209203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 630-22-8 | C5H11NS | 117.21 g/mol

2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 630-22-8
Molecular Formula C5H11NS
Molecular Weight (g/mol) 117.21
MDL Number MFCD09742834
InChI Key FJZJUSOFGBXHCV-UHFFFAOYSA-N
Synonym 2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio
PubChem CID 3031130
IUPAC Name 2,2-dimethylpropanethioamide
SMILES CC(C)(C)C(=S)N

Specifications

Melting Point 113°C to 117°C
Quantity 1 g
Solubility Information Soluble in water (10g/L) (25°C).
Formula Weight 117.21
Percent Purity 97%
Chemical Name or Material 2,2,2-Trimethylthioacetamide
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only