missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylacetophenone, 97%

Catalog No. AAL0872914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0872914 25 g
AAL0872903 1 g
AAL0872906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0872914 Supplier Thermo Scientific Chemicals Supplier No. L0872914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 938-16-9 | C11H14O | 162.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.

Chemical Identifiers

CAS 938-16-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00008844
InChI Key OECPUBRNDKXFDX-UHFFFAOYSA-N
Synonym 2,2-dimethylpropiophenone, pivalophenone, 2,2,2-trimethylacetophenone, phenyl tert-butyl ketone, tert-butyl phenyl ketone, 1-propanone, 2,2-dimethyl-1-phenyl, unii-96ba178unx, t-bubz, t-butyl phenyl ketone, dsstox_cid_28184
PubChem CID 70308
IUPAC Name 2,2-dimethyl-1-phenylpropan-1-one
SMILES CC(C)(C)C(=O)C1=CC=CC=C1

Specifications

Density 0.97
Boiling Point 219°C to 222°C
Flash Point 87°C (188°F)
Refractive Index 1.5084
Quantity 25 g
Beilstein 1906460
Solubility Information Not miscible with water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2,2,2-Trimethylacetophenone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-338-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only