Learn More
2,2,2-Trifluoroethylamine hydrochloride, 98%
CAS: 373-88-6 | C2H5ClF3N | 135.51 g/mol
Supplier: Thermo Scientific Chemicals A1490418
Description
2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff
Solubility
It is Soluble in 0.7 to water, but soluble in ethanol and chloroform, slightly soluble in benzene, hardly soluble in ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away from strong oxidizing agents, acids, acid chlorides, acid anhydrides.
Specifications
| 2,2,2-Trifluoroethylamine hydrochloride | |
| 218°C (sublimation) | |
| CF3CH2NH2·HCl | |
| 50 g | |
| Hygroscopic | |
| It is Soluble in 0.7 to water,but soluble in ethanol and chloroform,slightly soluble in benzene,hardly soluble in ether. | |
| [H+].[Cl-].NCC(F)(F)F | |
| 135.51 | |
| 135.52 |
| 373-88-6 | |
| C2H5ClF3N | |
| MFCD00012875 | |
| 3652103 | |
| 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1 | |
| ZTUJDPKOHPKRMO-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethanamine;hydrochloride | |
| 9772 | |
| 98% |
Chemical Identifiers
| 373-88-6 | |
| 135.51 | |
| ZTUJDPKOHPKRMO-UHFFFAOYSA-N | |
| 9772 | |
| [H+].[Cl-].NCC(F)(F)F |
| C2H5ClF3N | |
| MFCD00012875 | |
| 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1 | |
| 2,2,2-trifluoroethanamine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 206-771-1
RTECSNumber : KS0250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only